Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (INN-hFSH-132), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062251IR
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Chorionic Gonadotropin alpha Chain (hCG alpha) | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CGA | |
| Human Follicle Stimulating Hormone (FSH) | |
| 0.25 mL | |
| Primary | |
| A high affinity antibody recognising the alpha 4 epitope of the alpha-subunit of Human Chorionic Gonadotrophin (hCG), hLH, hFSH, hTSH and free alpha-subunit. The recognised epitope comprises amino-acids 15-22 of the human sequence. This antibody will not cross-react with glycoprotein hormones of various animal species. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| INN-hFSH-132 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 1081 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction