Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (INN-hFSH-158), Alexa Fluor™ 700

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062226AF700
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Western Blot, ELISA.Specifications
Chorionic Gonadotropin alpha Chain (hCG alpha) | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CGA | |
Human FSH | |
0.25 mL | |
Primary | |
This antibody reacts with the alpha 5 epitope on the alpha subunit of hCG, hLH, hFSH, hISH and the free alpha subunit. The alpha 5 epitope is destroyed upon iodination. | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
INN-hFSH-158 | |
Western Blot, ELISA | |
CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA | |
Mouse | |
Protein A or G purified | |
RUO | |
1081 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction