Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (27E8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108025
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Sandwich ELISA.Specifications
Chorionic Gonadotropin beta Chain (HCG beta) | |
Monoclonal | |
Unconjugated | |
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This specifically recognizes beta subunit of hCG. It does not cross-react with hLH, hTSH, hFSH. | |
Store at 4C. Do not freeze. | |
IgG1 |
ELISA | |
2700000000 | |
Sandwich ELISA 1:100-1:2000 | |
CGB3 | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human HCG (whole molecule). | |
0.2 mg | |
Cancer | |
1082 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction