Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin beta Chain (hCG beta) Antibody (HCGa/7806R), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$247.50 - $474.50

Specifications

Antigen Chorionic Gonadotropin beta Chain (hCG beta)
Clone HCGa/7806R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB234489
SDP
View Documents
Novus Biologicals
NBP320621100UG
100 μg
Each for $474.50
Only null left
Add to Cart
 
NB235298
SDP
View Documents
Novus Biologicals
NBP32062120UG
20 μg
Each for $247.50
Only null left
Add to Cart
 
Description

Description

Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal antibody specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Chorionic Gonadotropin beta Chain (hCG beta)
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer
10mM PBS with 0.05% BSA
1082
IgG κ
Protein A or G purified
HCGa/7806R
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB
Recombinant fragment (around aa1-116) of human Chorionic Gonadotropin beta Chain (hCG beta) protein (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.