Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-45), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062225UV
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for ELISA.Specifications
hCG holo C3 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CGB3 | |
Human hCG holo C3 | |
0.25 mL | |
Cancer | |
1082 | |
Human | |
Purified |
ELISA | |
INN-hCG-45 | |
ELISA | |
CGB chorionic gonadotropin, beta polypeptide, CGB3, CGB5, CGB7, CGB8, hCGB | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This antibody reacts with hCG holo C3 recognising the c3 epitope on hCG. No subunit detection is observed. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction