Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-53), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063167H
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA.Specifications
| hCG holo C2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CGB3 | |
| Made to Human hCG holo C2 | |
| 0.25 mL | |
| Cancer | |
| 1082 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| INN-hCG-53 | |
| Western Blot, ELISA | |
| choriogonadotropin subunit beta, CGB3, CGB5, CGB7, CGB8, chorionic gonadotropin, beta polypeptide, hCGB | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| NB100-63167 recognizes epitope c2 on hCG holo C2 . It does not recognize free hCG subunits. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction