Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (SPM529), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234416UV
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Chorionic Gonadotropin beta Chain (HCG beta) | |
Monoclonal | |
DyLight 350 | |
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB | |
Mouse | |
22 kDa | |
0.1 mL | |
Cancer | |
1082 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
SPM529 | |
Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
CGB3 | |
Recombinant Chorionic Gonadotropin beta Chain (hCG beta) protein (Uniprot: P01233) | |
Protein A or G purified | |
RUO | |
Primary | |
This monoclonal antibody reacts with a protein of 22kDa, identified as beta sub-unit of HCG. It does not cross react with the alpha sub-unit. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the subunit is identical to that of thyroid stimulating hormone (TSH) follicle stimulating hormone (FSH), and luteinizing hormone (LH). hCG monoclonal antibody detects cells and tumors of trophoblastic origin such as choriocarcinoma. Large cell carcinoma and adenocarcinoma of the lung demonstrate anti-hCG positivity in 90% and 60% of cases respectively. 20% of lung squamous cell carcinomas are positive. hCG expression by non-trophoblastic tumors may indicate aggressive behavior. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction