Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin Chicken anti-Human, Polyclonal , Abnova™

Catalog No. 89933972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-972 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-972 Supplier Abnova Corporation Supplier No. PAB29776

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chicken polyclonal antibody raised against native human Chorionic Gonadotropin.

Specifications

Antigen Chorionic Gonadotropin
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Chicken
Immunogen Native purified human Chorionic Gonadotropin.
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.