Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Chorionic Gonadotropin, Human/hCG Ab-2, Rabbit Polyclonal Antibody

Catalog No. RB059R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RB059R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RB059R1 Supplier Epredia™ Supplier No. RB059R1

Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ Chorionic Gonadotropin, Human/hCG Ab-2, Rabbit Polyclonal Antibody.

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha- and Beta-subunits. The alpha-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin-fixed)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Purified Beta-subunit of HCG
Quantity 1 mL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.