Learn More
CHP1 Mouse anti-Human, Clone: OTI6F5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810173

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a phosphoprotein that binds to the Na+/H+ exchanger NHE1. This protein serves as an essential cofactor which supports the physiological activity of NHE family members and may play a role in the mitogenic regulation of NHE1. The protein shares similarity with calcineurin B and calmodulin and it is also known to be an endogenous inhibitor of calcineurin activity.Specifications
CHP1 | |
Monoclonal | |
Unconjugated | |
CHP1 | |
CHP, SLC9A1BP, Sid470p, p22, p24 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11261 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6F5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q99653 | |
CHP1 | |
Full length human recombit protein of human CHP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.