Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582255 Supplier Invitrogen™ Supplier No. PA582255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFSHSQITRL YSRFTSLDKG ENGTLSREDF QRIPELAINP LGDRIINAFF PEGEDQVNFR GFMRTLAHFR PIEDNEKSKD VNGPEPLNSR SNKLHFAFRL YDLDKDEKIS RDELLQVLRM MVGVNIS Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Regulates centrosome duplication, probably by inhibiting the kinase activity of ROCK2. Proposed to act as co-chaperone for HSP90. May play a role in the regulation of NOD1 via a HSP90 chaperone complex. In vitro, has intrinsic chaperone activity. This function may be achieved by inhibiting association of ROCK2 with NPM1. Involved in stress response. Prevents tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHP1
Gene Accession No. Q99653
Gene Alias 1500003O03Rik; AA960066; AI046351; Cahp; calcineurin B homolog; calcineurin B homologous protein 1; Calcineurin B-like protein; calcineurin homologous protein; calcineurin like EF-hand protein 1; calcineurin-like EF hand protein 1; calcineurin-like EF-hand protein 1; calcium binding protein P22; calcium-binding protein CHP; Calcium-binding protein p22; CHP; chp1; Chromo domain-containing protein 1; chromodomain protein Chp1; EF-hand Ca2+-binding protein p22; EF-hand calcium-binding domain-containing protein p22; p22; p24; si:dkey-41n20.1; sid 470; Sid470; Sid470p; SLC9A1 binding protein; SLC9A1BP; SPAC18G6.02c; Unknown (protein for MGC:127971); Unknown (protein for MGC:63904); vac; Wrch2; wu:fc69b04; zgc:63904
Gene Symbols CHP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHP1. Recombinant protein control fragment (Product #RP-89506).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11261
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.