Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHPT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523695
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523695 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523695 Supplier Invitrogen™ Supplier No. PA523695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine and mouse based on sequence homology.

Catalyzes the final step of de novo phosphatidylcholine (PC) synthesis, i.e. the transfer of choline phosphate from CDP-choline to the free hydroxyl of a diacylglycerol (DAG), producing a PC. It thereby plays a central role in the formation and maintenance of vesicular membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHPT1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CHPT1
Gene Accession No. Q8WUD6
Gene Alias AAPT1-like protein; choline phosphotransferase 1; cholinephosphotransferase 1; cholinephosphotransferase 1 alpha; Chpt1; CPT; CPT1; Diabetic nephropathy-associated gene transcript 1; diacylglycerol cholinephosphotransferase 1; Dn4; hCPT1; mCHPT1; MSTP022; phosphatidylcholine synthesizing enzyme
Gene Symbols CHPT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 7-33 amino acids from the N-terminal region of human CHPT1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.