Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRAC1 Monoclonal Antibody (4B8)
GREENER_CHOICE

Catalog No. PIMA522014
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522014 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522014 Supplier Invitrogen™ Supplier No. MA522014
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MADVVVGKDK GGEQRLISLP LSRIRVIMKS SPEVSSINQE ALVLTAKATE LFVQCLATYS YRHGSGKEKK VLTYSDLANT AQQSETFQFL ADILPKKILA SKYLKMLKEE KREEDEENDN DNESDHDEAD S.

CHRAC1 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.[supplied by OMIM, Apr 2004].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRAC1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CHRAC1
Gene Accession No. Q9NRG0
Gene Alias 2410152E03Rik; 2810406L04Rik; CHARC1; CHARC15; CHRAC1; CHRAC-1; CHRAC15; CHRAC-15; CHRC1; chromatin accessibility complex 1; chromatin accessibility complex 15 kDa protein; Chromatin accessibility complex protein 1; DNA polymerase epsilon subunit p15; histone-fold protein CHRAC15; HuCHRAC15; NF-YC-like protein; YCL1; YC-like protein 1
Gene Symbols CHRAC1
Host Species Mouse
Immunogen CHRAC1 (AAH15891, 1 a.a. ~ 131 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54108
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.