Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHRDL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551368
Description
Immunogen sequence: DKKYRVGERW HPYLEPYGLV YCVNCICSEN GNVLCSRVRC PNVHCLSPVH IPHLCCPRCP EDSLPPVNNK VTSKSCEYNG TTYQHGELFV AEGLFQNRQP NQCTQCSCSE GNVYC Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
CHRDL1 is an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
CHRDL1 | |
Polyclonal | |
Unconjugated | |
CHRDL1 | |
CHL; CHL1; chordin like 1; chordin-like 1; chordin-like protein 1; Chrdl1; dA141H5.1; Kjr; Kohjirin; MGC1; MGCN; neuralin 1; neuralin-1; neurogenesin 1; Neurogenesin-1; Ng1; Nrln1; Ventroptin; ventroptin-alpha; ventroptin-beta; VOPT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BU40 | |
CHRDL1 | |
Recombinant protein corresponding to Human CHRDL1. Recombinant protein control fragment (Product #RP-91922). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction