Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRDL2 Monoclonal Antibody (272905)
GREENER_CHOICE

Catalog No. PIMA523973
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523973 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523973 Supplier Invitrogen™ Supplier No. MA523973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blots, no cross-reactivity with recombinant mouse CHRDL1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

CHRDL2 encodes a member of the chordin family of proteins. Chordin family members are secreted proteins that share a cysteine-rich pro-collagen repeat domain and associate with members of the transforming growth factor beta superfamily. In vitro assays demonstrate a direct interaction between the encoded protein and human activin A. This gene is expressed in many tissues including osteoblasts, where it is differentially expressed during differentiation. In addition, its expression is upregulated in human osteoarthritic joint cartilage, suggesting a role in adult cartilage regeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRDL2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 272905
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Chrdl2
Gene Accession No. Q6WN34
Gene Alias 1810022C01Rik; BNF1; BNF-1; Breast tumor novel factor 1; CHL2; chordin like 2; chordin-like 2; chordin-like protein 2; Chordin-related protein 2; CHRDL2; FKSG37; UNQ765/PRO1557
Gene Symbols Chrdl2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chordin-like 2/CHRDL2 Arg28-Thr451.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25884
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.