Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHREBP Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 116 publications
$269.00 - $572.50
Specifications
Antigen | CHREBP |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21282101), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 26181104), Gel Super Shift Assays reported in scientific literature (PMID 20025850), SDS-Page reported in scientific literature (PMID 35041621), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31668386) |
Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400135SS
![]() |
Novus Biologicals
NB400135SS |
0.025 mL |
Each for $269.00
|
|
|||||
NB400135
![]() |
Novus Biologicals
NB400135 |
0.1 mL |
Each for $572.50
|
|
|||||
Description
CHREBP Polyclonal specifically detects CHREBP in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays, Knockout Validated.Specifications
CHREBP | |
Western Blot 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21282101), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 26181104), Gel Super Shift Assays reported in scientific literature (PMID 20025850), SDS-Page reported in scientific literature (PMID 35041621), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31668386) | |
Polyclonal | |
Rabbit | |
Cancer, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein | |
MLXIPL | |
IgG | |
Affinity Purified | |
Human, rat and mouse (PMID 21835137). |
1 mg/mL | |
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Q9NP71 | |
51085 | |
A C-terminal synthetic peptide made to the human CHREBP protein sequence (between residues 800-852). [UniProt# Q9NP71, Isoform 1/Alpha] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
95 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title