Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHREBP Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400136H
Description
CHREBP Polyclonal specifically detects CHREBP in Human, Mouse samples. It is validated for Western Blot.Specifications
CHREBP | |
Polyclonal | |
Western Blot | |
Q9NP71 | |
MLXIPL | |
An internal synthetic peptide made to the human CHREBP protein sequence (between residues 50-150). [UniProt# Q9NP71, Isoform 1/Alpha] | |
0.1 mL | |
Cancer, Chromatin Research, Lipid and Metabolism, Transcription Factors and Regulators | |
51085 | |
Human, Mouse | |
IgG |
Western Blot | |
HRP | |
PBS with No Preservative | |
BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and Mouse. Immunogen sequence has 87% identity with rat. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction