Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHREBP Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 6 publications

$206.50 - $499.50

Specifications

Antigen CHREBP
Concentration 1 mg/mL
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB400136SS
SDP
View Documents
Novus Biologicals
NB400136SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB400136
SDP
View Documents
Novus Biologicals
NB400136
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

CHREBP Polyclonal specifically detects CHREBP in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CHREBP
Western Blot, Immunocytochemistry/Immunofluorescence 1:100
Polyclonal
Rabbit
Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.01% Sodium Azide
BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein
MLXIPL
IgG
Affinity Purified
Human and Mouse. Immunogen sequence has 87% identity with rat.
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse
Q9NP71
51085
An internal synthetic peptide made to the human CHREBP protein sequence (between residues 50-150). [UniProt# Q9NP71, Isoform 1/Alpha]
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.