Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ChREBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584659 Supplier Invitrogen™ Supplier No. PA584659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSERRLSGDL SSMPGPGTLS VRVSPPQPIL SRGRPDSNKT ENRRITHISA E Highest antigen sequence indentity to the following orthologs: Rat - 35%, Mouse - 75%.

ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ChREBP
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MLXIPL
Gene Accession No. Q9NP71
Gene Alias BHLHD14; carbohydrate response element binding protein; carbohydrate-responsive element-binding protein; ChREBP; Class D basic helix-loop-helix protein 14; MIO; MLX interacting protein like; MLX interacting protein-like; MLX interacting protein-like beta; Mlx interactor; MLX-interacting protein-like; MLXIPL; MONDOB; putative hepatic transcription factor; WBSCR14; Williams Beuren syndrome chromosome region 14; williams-Beuren syndrome chromosomal region 14 protein; Williams-Beuren syndrome chromosomal region 14 protein homolog; Williams-Beuren syndrome chromosome region 14 homolog; Williams-Beuren syndrome chromosome region 14 protein 1; Williams-Beuren syndrome chromosome region 14 protein 2; Williams-Beuren syndrome chromosome region 14 protein 3; WS basic-helix-loop-helix leucine zipper protein; WS-bHLH
Gene Symbols MLXIPL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MLXIPL. Recombinant protein control fragment (Product #RP-105326).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.