Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHRFAM7A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215894
Description
CHRFAM7A Polyclonal specifically detects CHRFAM7A in Human samples. It is validated for Western Blot.Specifications
CHRFAM7A | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
CHRNA7, CHRNA7 (cholinergic receptor, nicotinic, alpha 7, exons 5-10) and FAM7A (familywith sequence similarity 7A, exons A-E) fusion, CHRNA7 (cholinergic receptor, nicotinic, alpha polypeptide 7, exons 5-10) andFAM7A (family with sequence similarity 7A, exons A-E) fusion, CHRNA7-DR1alpha 7 neuronal nicotinic acetylcholine receptor-FAM7A hybrid, CHRNA7-FAM7A fusion protein, D-10alpha-7 nicotinic cholinergic receptor subunit, MGC120482, MGC120483 | |
Rabbit | |
46 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
0.56mg/mL | |
Western Blot 1:500-1:3000 | |
Q494W8 | |
CHRFAM7A | |
Recombinant protein encompassing a sequence within the center region of human CHRFAM7A. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
89832 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction