Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545173 Supplier Invitrogen™ Supplier No. PA545173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VKRPTKKGRD RAGKGQKPRG KEQLAKRKTF SLVKEKKAAR TLSAILLAFI Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 1 is involved in mediation of vagally-induced bronchoconstriction and in the acid secretion of the gastrointestinal tract. The gene encoding this receptor is localized to 11q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHRM1
Gene Accession No. P11229
Gene Alias acetylcholine receptor; acetylcholine receptor, muscarinic 1; AChR M1; cholinergic receptor; cholinergic receptor muscarinic 1; Cholinergic receptor, muscarin 1; cholinergic receptor, muscarinic 1; cholinergic receptor, muscarinic 1, CNS; CHRM1; Chrm-1; HM1; M1; M1 mAChR; m1 muscarinic acetylcholine receptor protein; M1R; MGC30125; muscarinic acetylcholine receptor M1; muscarinic receptor M1
Gene Symbols CHRM1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CHRM1 (aa 326-375).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1128
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.