Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHRM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586494
Description
Purity is > 95% by SDS-PAGE.
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 1 is involved in mediation of vagally-induced bronchoconstriction and in the acid secretion of the gastrointestinal tract. The gene encoding this receptor is localized to 11q13.
Specifications
CHRM1 | |
Polyclonal | |
Unconjugated | |
CHRM1 | |
acetylcholine receptor; acetylcholine receptor, muscarinic 1; AChR M1; cholinergic receptor; cholinergic receptor muscarinic 1; Cholinergic receptor, muscarin 1; cholinergic receptor, muscarinic 1; cholinergic receptor, muscarinic 1, CNS; CHRM1; Chrm-1; HM1; M1; M1 mAChR; m1 muscarinic acetylcholine receptor protein; M1R; MGC30125; muscarinic acetylcholine receptor M1; muscarinic receptor M1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1128, 12669, 25229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P08482, P11229, P12657 | |
CHRM1 | |
Synthetic peptide, corresponding to amino acids 311-360 of Human mAChR M1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction