Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRM2 Monoclonal Antibody (B8E5)
GREENER_CHOICE

Catalog No. FERMA549029
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549029 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549029 Supplier Invitrogen™ Supplier No. MA549029
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody B8E5 acts by functional dimerization of the receptor resulting in stabilization of the constitutive active receptor dimers and paradoxically induces a small decrease in carbachol affinity for the M2 receptor. It recognizes the pentapeptide VRTVE (aa 168-172) corresponding to the N-terminal part of the second extracellular loop of the human M2 receptor.

Muscarinic cholinergic receptor 2 (CHRM2) is an Acetylcholine Receptor has been suggested to function as a presynaptic autoreceptor that inhibits acetylcholine release. The M2 receptor has also been suggested to mediate smooth muscle contractility. Alzheimer's disease (AD) involves selective loss of M2 subtype receptors in cortical and hippocampal regions of the human brain. Increase in M2 receptor heterozygosity has been associated with depression in women. The M2 receptor has been reported to be expressed in various regions of the brain including the cerebellum, cerebral cortex, hippocampus, medulla, striatum, and thalamus, and in lung and prostate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRM2
Applications Functional Assay, Inhibition Assays, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone B8E5
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene CHRM2
Gene Accession No. P08172
Gene Alias 7TM receptor; acetylcholine receptor; acetylcholine receptor, muscarinic 2; AChR M2; AChR-M2; Acm2; cardiac muscarinic acetylcholine receptor; cholinergic receptor; cholinergic receptor muscarinic 2; cholinergic receptor, muscarinic 2; cholinergic receptor, muscarinic 2, cardiac; cholinergic receptor, muscarinic 2, isoform a; Chrm2; Chrm-2; FLJ43243; HM2; M2; M2 muscarinic acetylcholine receptor; m2AChR; M2-mAChR; MGC120006; MGC120007; muscarinic acetylcholine receptor 2; muscarinic acetylcholine receptor m2; muscarinic M2 receptor; muscarinic receptor m2; unnamed protein product
Gene Symbols CHRM2
Host Species Mouse
Immunogen Peptide VRTVEDGECYIQFFSNAAVTFGTAI(C): Human second extracellular loop residues 168-192.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1129
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.