Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590608 Supplier Invitrogen™ Supplier No. PA590608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HISRASKSRI KKDKKEPVAN QDPVSPSLVQ GRIVKPNNNN MPSSDDGLEH NKIQNGKAPR DPVTENCVQG EEKESSNDST SVSAVASNMR DDEITQDENT VSTSLGHSKD ENSKQTCIRI GTKTPKSDSC TPTNTTVEVV GSSGQNGDEK QNIVARKIVK MTKQPAKKKP PPSREKKVTR T; Positive Samples: SH-SY5Y, HepG2; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.

Muscarinic cholinergic receptor 2 (CHRM2) is an Acetylcholine Receptor has been suggested to function as a presynaptic autoreceptor that inhibits acetylcholine release. The M2 receptor has also been suggested to mediate smooth muscle contractility. Alzheimer's disease (AD) involves selective loss of M2 subtype receptors in cortical and hippocampal regions of the human brain. Increase in M2 receptor heterozygosity has been associated with depression in women. The M2 receptor has been reported to be expressed in various regions of the brain including the cerebellum, cerebral cortex, hippocampus, medulla, striatum, and thalamus, and in lung and prostate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRM2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHRM2
Gene Accession No. P08172
Gene Alias 7TM receptor; acetylcholine receptor; acetylcholine receptor, muscarinic 2; AChR M2; AChR-M2; Acm2; cardiac muscarinic acetylcholine receptor; cholinergic receptor; cholinergic receptor muscarinic 2; cholinergic receptor, muscarinic 2; cholinergic receptor, muscarinic 2, cardiac; cholinergic receptor, muscarinic 2, isoform a; Chrm2; Chrm-2; FLJ43243; HM2; M2; M2 muscarinic acetylcholine receptor; m2AChR; M2-mAChR; MGC120006; MGC120007; muscarinic acetylcholine receptor 2; muscarinic acetylcholine receptor m2; muscarinic M2 receptor; muscarinic receptor m2; unnamed protein product
Gene Symbols CHRM2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 208-388 of human Muscarinic AChR M2 (ACM2) (NP_0010066291).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1129
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.