Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRM3 Monoclonal Antibody (4A6B7)
GREENER_CHOICE

Catalog No. PIMA538496
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538496 Supplier Invitrogen™ Supplier No. MA538496
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRM3
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4A6B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CHRM3
Gene Accession No. P20309
Gene Alias acetylcholine receptor; acetylcholine receptor, muscarinic 3; AChR M3; cholinergic receptor; cholinergic receptor muscarinic 3; cholinergic receptor, muscarinic 3; cholinergic receptor, muscarinic 3, cardiac; CHRM3; Chrm-3; EGBRS; HM3; M3; m3 muscarinic receptor; M3R; Mm3 mAChR; muscarinic acetylcholine receptor 3; muscarinic acetylcholine receptor M3; PBS
Gene Symbols CHRM3
Host Species Mouse
Immunogen Purified recombinant fragment of human CHRM3 (aa: extra mix) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.