Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrm3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982453
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2453 Supplier Bioss Supplier No. BS1289R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue.

Specifications

Antigen Chrm3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CHRM3
Gene Accession No. P20309
Gene Alias acetylcholine receptor, muscarinic 3; AChR M3; cholinergic receptor muscarinic 3; cholinergic receptor, muscarinic 3; cholinergic receptor, muscarinic 3, cardiac; Chrm3; Chrm-3; EGBRS; HM3; M3; m3 muscarinic receptor; M3R; Mm3 mAChR; muscarinic acetylcholine receptor 3; Muscarinic acetylcholine receptor M3; PBS
Gene Symbols CHRM3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Chrm3 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1131
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.