Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567650 Supplier Invitrogen™ Supplier No. PA567650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Muscarinic acetylcholine receptor M4 is an Acetylcholine Receptor. Unlike muscarinic receptor M2, the M4 receptor is not involved in smooth muscle contraction. The clinical relevance of this receptor is unknown. However, mouse studies have linked it to adenylyl cyclase inhibition. Muscarinic acetylcholine receptor M4 has been reported to be expressed in many regions of the rat brain, particularly the striatum and hippocampus. ESTs have been isolated from human breast and eye libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRM4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Gene CHRM4
Gene Accession No. P08173, P32211
Gene Alias acetylcholine receptor; acetylcholine receptor, muscarinic 4; AChR M4; cholinergic receptor; cholinergic receptor muscarinic 4; cholinergic receptor, muscarin 4; cholinergic receptor, muscarinic 4; CHRM4; Chrm-4; HM4; M4; M4R; Mm4 mAChR; muscarinic acetylcholine receptor 4; muscarinic acetylcholine receptor M4
Gene Symbols CHRM4
Host Species Rabbit
Immunogen Synthesized peptide derived from human CHRM4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1132, 12672
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.