Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRM4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA123544
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA123544 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA123544 Supplier Thermo Scientific Supplier No. PA123544

Rabbit Polyclonal Antibody

PA1-23544 detects Muscarinic Acetylcholine Receptor M4 in Human samples.

Muscarinic acetylcholine receptor M4 is an Acetylcholine Receptor. Unlike muscarinic receptor M2, the M4 receptor is not involved in smooth muscle contraction. The clinical relevance of this receptor is unknown. However, mouse studies have linked it to adenylyl cyclase inhibition. Muscarinic acetylcholine receptor M4 has been reported to be expressed in many regions of the rat brain, particularly the striatum and hippocampus. ESTs have been isolated from human breast and eye libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRM4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene CHRM4
Gene Accession No. P08173
Gene Alias acetylcholine receptor; acetylcholine receptor, muscarinic 4; AChR M4; cholinergic receptor; cholinergic receptor muscarinic 4; cholinergic receptor, muscarin 4; cholinergic receptor, muscarinic 4; CHRM4; Chrm-4; HM4; M4; M4R; Mm4 mAChR; muscarinic acetylcholine receptor 4; muscarinic acetylcholine receptor M4
Gene Symbols CHRM4
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human mAChR M4
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.