Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109537 Supplier Invitrogen™ Supplier No. PA5109537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRSTSTTGKP SQATGPSANW AKAEQLTTCS SYPSSEDEDK PATDPVLQVV YKSQGKESPG EEFSAEETEE TFVKAETEKS DYDTPNYLLS PAAAHR.

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The clinical implications of this receptor are unknown; however, stimulation of this receptor is known to increase cyclic AMP levels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRM5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CHRM5
Gene Accession No. P08911, P08912, Q920H4
Gene Alias acetylcholine receptor; acetylcholine receptor, muscarinic 5; AChR-M5; cholinergic receptor; cholinergic receptor muscarinic 5; cholinergic receptor, muscarinic 5; Chrm5; Chrm-5; HM5; m5; M5R; muscarinic acetylcholine receptor 5; muscarinic acetylcholine receptor M5
Gene Symbols CHRM5
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1133, 213788, 53949
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.