Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNA10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550074
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550074 Supplier Invitrogen™ Supplier No. PA550074
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CHRNA10 protein.

Ionotropic receptor with a probable role in the modulation of auditory stimuli. Agonist binding may induce an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane. The channel is permeable to a range of divalent cations including calcium, the influx of which may activate a potassium current which hyperpolarizes the cell membrane. In the ear, this may lead to a reduction in basilar membrane motion, altering the activity of auditory nerve fibers and reducing the range of dynamic hearing. This may protect against acoustic trauma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHRNA10
Gene Accession No. Q9GZZ6
Gene Alias acetylcholine receptor; acetylcholine receptor, nicotinic, alpha 10 (neuronal); cholinergic receptor; cholinergic receptor nicotinic alpha 10 subunit; cholinergic receptor, nicotinic alpha 10; cholinergic receptor, nicotinic, alpha 10 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 10; Chrna10; NACHR alpha 10; NACHR alpha-10; NACHRA10; Neuronal acetylcholine receptor subunit alpha-10; neuronal nicotinic acetylcholine receptor subunit; nicotinic acetylcholine receptor subunit alpha 10; nicotinic acetylcholine receptor subunit alpha-10
Gene Symbols CHRNA10
Host Species Rabbit
Immunogen Synthesized peptide derived from C-terminal of human CHRNA10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 504186, 57053
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.