Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNA2 Mouse anti-Human, Clone: 5B2.1, MilliporeSigma™

Catalog No. MABN224925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2249-25 25 μg
MABN2249-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2249-25 Supplier MilliporeSigma Supplier No. MABN224925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CHRNA2, clone 5B2.1 Antibody is a highly specific mouse monoclonal antibody that targets Neuronal acetylcholine receptor subunit alpha-2 (CHRNA2) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5B2.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CHRNA2
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 52 amino acids from the N-terminal region of human Neuronal acetylcholine receptor subunit alpha-2 (CHRNA2).
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000733
Test Specificity Clone 5B2.1 detects human Neuronal acetylcholine receptor subunit alpha-2 (CHRNA2). It targets an epitope within 52 amino acids from the N-terminal region extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.