Learn More
Invitrogen™ CHRNA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595837
Description
Immunogen sequence: SEAEHRLFER LFEDYNEIIR PVANVSDPVI IHFEVSMSQL VKVDEVNQIM ETNLWLKQIW NDYKLKWNPS DYGGAEFMRV PAQKIWKPDI VLYNNAVGDF QVDDKTKALL KYTGEVTWIP PAIFKSSCKI DVTYFPFDYQ NCTMKFGSWS YDKAKIDLVL IGSSMNLKDY WESGEWAIIK APGYKHDIKY NCCEEIYPDI TYSLYIRRL; Positive Samples: SH-SY5Y, HepG2, Mouse brain, Rat liver; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.
CHRNA3 locus encodes a member of the nicotinic acetylcholine receptor family of proteins. Members of this family of proteins form pentameric complexes comprised of both alpha and beta subunits. This locus encodes an alpha-type subunit, as it contains characteristic adjacent cysteine residues. The encoded protein is a ligand-gated ion channel that likely plays a role in neurotransmission. Polymorphisms in this gene have been associated with an increased risk of smoking initiation and an increased susceptibility to lung cancer. Alternatively spliced transcript variants have been described.
Specifications
CHRNA3 | |
Polyclonal | |
Unconjugated | |
CHRNA3 | |
(a)3; A730007P14Rik; acetylcholine receptor; Acetylcholine receptor alpha 3 (neuronal nicotine); Acra3; Acra-3; alpha 3; cetylcholine receptor alpha 3 (neuronal nicotine); cholinergic receptor; cholinergic receptor nicotinic alpha 3 subunit; cholinergic receptor nicotinic alpha polypeptide 4; cholinergic receptor, nicotinic alpha 3; cholinergic receptor, nicotinic, alpha 3 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 3; CHRNA3; LNCR2; nAChR alpha 3; NACHRA3; Neuronal acetylcholine receptor subunit alpha-3; neuronal nicotinic acetylcholine receptor, alpha 3 subunit; neuronal nicotinic acetylcholine receptor, alpha3 subunit; PAOD2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
110834, 1136, 25101 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04757, P32297, Q8R4G9 | |
CHRNA3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 32-240 of human CHRNA3 (NP_0007342). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.