Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNA3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987273
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7273 Supplier Bioss Supplier No. BS6455R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHRNA3 locus encodes a member of the nicotinic acetylcholine receptor family of proteins. Members of this family of proteins form pentameric complexes comprised of both alpha and beta subunits. This locus encodes an alpha-type subunit, as it contains characteristic adjacent cysteine residues. The encoded protein is a ligand-gated ion channel that likely plays a role in neurotransmission. Polymorphisms in this gene have been associated with an increased risk of smoking initiation and an increased susceptibility to lung cancer. Alternatively spliced transcript variants have been described.

Specifications

Antigen CHRNA3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CHRNA3
Gene Accession No. P32297
Gene Alias (a)3; A730007P14Rik; Acetylcholine receptor alpha 3 (neuronal nicotine); Acra3; Acra-3; alpha 3; cetylcholine receptor alpha 3 (neuronal nicotine); cholinergic receptor nicotinic alpha 3 subunit; cholinergic receptor nicotinic alpha polypeptide 4; cholinergic receptor, nicotinic alpha 3; cholinergic receptor, nicotinic, alpha 3 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 3; Chrna3; LNCR2; nAChR alpha 3; NACHRA3; Neuronal acetylcholine receptor subunit alpha-3; neuronal nicotinic acetylcholine receptor, alpha 3 subunit; neuronal nicotinic acetylcholine receptor, alpha3 subunit; PAOD2
Gene Symbols CHRNA3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CHRNA3/AChR3 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1136
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.