Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNA4 Mouse anti-Human, Clone:7B4H1, Abnova™

Catalog No. 89334005 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-005 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-005 Supplier Abnova Corporation Supplier No. MAB17539
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CHRNA4.

This gene encodes a nicotinic acetylcholine receptor, which belongs to a superfamily of ligand-gated ion channels that play a role in fast signal transmission at synapses. These pentameric receptors can bind acetylcholine, which causes an extensive change in conformation that leads to the opening of an ion-conducting channel across the plasma membrane. This protein is an integral membrane receptor subunit that can interact with either nAChR beta-2 or nAChR beta-4 to form a functional receptor. Mutations in this gene cause nocturnal frontal lobe epilepsy type 1. Polymorphisms in this gene that provide protection against nicotine addiction have been described. [provided by RefSeq]

Specifications

Antigen CHRNA4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7B4H1
Conjugate Unconjugated
Description cholinergic receptor, nicotinic, alpha 4
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CHRNA4
Gene Alias BFNC, EBN, EBN1, FLJ95812, NACHR, NACHRA4, NACRA4
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 29-242 of human CHRNA4 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 1137
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.