Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534714 Supplier Invitrogen™ Supplier No. PA534714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (97%), Rat (96%), Chicken (88%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a nicotinic acetylcholine receptor, which belongs to a superfamily of ligand-gated ion channels that play a role in fast signal transmission at synapses. These pentameric receptors can bind acetylcholine, which causes an extensive change in conformation that leads to the opening of an ion-conducting channel across the plasma membrane. This protein is an integral membrane receptor subunit that can interact with either nAChR beta-2 or nAChR beta-4 to form a functional receptor. Mutations in this gene cause nocturnal frontal lobe epilepsy type 1. Polymorphisms in this gene that provide protection against nicotine addiction have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA4
Applications Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CHRNA4
Gene Accession No. P43681
Gene Alias a4 nicotinic receptor; acetylcholine receptor; acetylcholine receptor alpha 4 neural; acetylcholine receptor, neuronal, nicotinic, alpha-4 subunit; Acra4; Acra-4; alpha4 nAChR; alpha4-nAChR; BFNC; cholinergic receptor; cholinergic receptor nicotinic alpha 4 subunit; cholinergic receptor, nicotinic alpha 4; cholinergic receptor, nicotinic, alpha 4 (neuronal); cholinergic receptor, nicotinic, alpha 4 subunit; cholinergic receptor, nicotinic, alpha polypeptide 4; CHRNA 4; Chrna4; EBN; EBN1; ENFL1; NACHR; nAChR alpha 4; NACHRA4; NACRA4; NARAC; neuronal acetylcholine receptor subunit alpha-4; Neuronal nicotinic acetylcholine receptor alpha 4 subunit; neuronal nicotinic acetylcholine receptor alpha-4 subunit
Gene Symbols CHRNA4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AChR alpha 4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.