Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHRNA5 Mouse anti-Human, Clone:2D2D10, Abnova™
Mouse monoclonal antibody raised against partial recombinant human CHRNA5.
Supplier: Abnova Corporation MAB17928
Description
Nicotinic acetylcholine receptors (nAChRs), such as CHRNA5, are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are thought to be (hetero)pentamers composed of homologous subunits. See MIM 118508 for additional background information on AChRs.[supplied by OMIM]Specifications
CHRNA5 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
CHRNA5 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
2D2D10 | |
cholinergic receptor, nicotinic, alpha 5 | |
In PBS (0.05% sodium azide) | |
LNCR2 | |
Recombinant protein corresponding to amino acids 23-254 of human CHRNA5. | |
1138 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction