Learn More
CHRNA5 Mouse anti-Human, Clone: OTI8H10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810073

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Nicotinic acetylcholine receptors (nAChRs), such as CHRNA5, are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are thought to be (hetero)pentamers composed of homologous subunits.Specifications
CHRNA5 | |
Monoclonal | |
Unconjugated | |
CHRNA5 | |
LNCR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1138 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8H10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P30532 | |
CHRNA5 | |
Human recombit protein fragment corresponding to amino acids 33-71 of human CHRNA5 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.