Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHRNA5 Mouse anti-Human, Mouse, Porcine, Clone: 3G8C11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CHRNA5 |
|---|---|
| Clone | 3G8C11 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6793
![]()
|
Proteintech Group Inc
663631IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Nicotinic acetylcholine receptors (nAChRs), such as CHRNA5, are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are thought to be (hetero)pentamers composed of homologous subunits.Specifications
| CHRNA5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CHRNA5, LNCR2, NACHRA5 | |
| CHRNA5 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 3G8C11 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig | |
| P30532, Q2MKA5 | |
| 100156023, 110835, 1138 | |
| CHRNA5 Fusion Protein Ag4447 | |
| Primary | |
| -20°C | |
| CHRNA5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title