Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562957 Supplier Invitrogen™ Supplier No. PA562957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQRGLSEPSS IAKHEDSLLK DLFQDYERWV RPVEHLNDK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Nicotinic acetylcholine receptors (nAChRs), such as CHRNA5, are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are thought to be (hetero)pentamers composed of homologous subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHRNA5
Gene Accession No. P30532
Gene Alias acetylcholine receptor; acetylcholine receptor alpha 5; acetylcholine receptor, nicotinic, alpha 5 (neuronal); Acra5; Acra-5; cholinergic receptor; cholinergic receptor nicotinic alpha 5 subunit; Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5; cholinergic receptor, nicotinic alpha 5; cholinergic receptor, nicotinic, alpha 5; cholinergic receptor, nicotinic, alpha 5 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 5; CHRNA5; hypothetical protein LOC550584; LNCR2; nAChR alpha 5; NACHRA5; Neuronal acetylcholine receptor subunit alpha-5; neuronal nicotinic acetylcholine receptor alpha5 subunit; neuronal nicotinic acetylcholine receptor, alpha5 subunit; nicotinic acetylcholine receptor alpha5 subunit; zgc:110642
Gene Symbols CHRNA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHRNA5. Recombinant protein control fragment (Product #RP-95689).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.