Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNA7 Monoclonal Antibody (7F10F1)
GREENER_CHOICE

Catalog No. PIMA531690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531690 Supplier Invitrogen™ Supplier No. MA531690
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31690 has been tested in indirect ELISA.

Chromosome 15 encodes over 700 genes and comprises nearly 3% of the human genome. CHRFAM7A, a fusion protein comprised of FAM7A and AChR alpha7, is encoded by the CHRFAM7A gene, which is located on a region of human chromosome 15 that is associated with several neuropsychiatric disorders, such as schizophrenia and bipolar affective disorder. CHRFAM7A, also known as D-10 or CHRNA7, is a 412 amino acid multi-pass membrane protein that belongs to the nicotinic acetylcholine receptor (nAChR) subfamily of ligand-gated ion channel proteins, a protein superfamily that mediates signal transmission at synapses. CHRFAM7A is expressed in the hippocampus. Alternative splicing of the CHRFAM7A gene produces two splice variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA7
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7F10F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CHRNA7
Gene Accession No. P36544, Q05941
Gene Alias a7 nicotinic acetylcholine receptor; acetylcholine receptor; acetylcholine receptor alpha 7 neural; Acra7; alpha 7 neuronal nicotinic acetylcholine receptor; alpha7; alpha-7 nicotinic cholinergic receptor subunit; alpha7 nicotinic receptor; alpha7-nAChR; BTX; bungarotoxin alpha; C holinergic receptor nicotinic alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); C holinergic receptor, nicotinic, alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); cholinergic receptor; cholinergic receptor nicotinic alpha 7 subunit; cholinergic receptor, nicotinic alpha 7; cholinergic receptor, nicotinic, alpha 7 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 7; CHRFAM7A; Chrna7; CHRNA7-2; CHRNA7-DR1; D-10; nAchR; nAChR alpha7; NACHRA7; NARAD; neuronal acetylcholine receptor protein, alpha-7 chain; neuronal acetylcholine receptor subunit alpha-7; neuronal nicotinic acetycholine receptor alpha 7; nicotinic receptor alpha 7 subunit
Gene Symbols CHRNA7
Host Species Mouse
Immunogen Purified recombinant fragment of human CHRNA7 (AA: extra 52-259) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1139, 25302
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.