Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHRNA9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546826
Description
Peptide sequence: MNWSHSCISF CWIYFAASRL RAAETADGKY AQKLFNDLFE DYSNALRPVE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Mouse: 93%; Rabbit: 100%; Zebrafish: 80%.
CHRNA9 is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor superfamily. CHRNA9 is a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. It is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea.This gene is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor gene superfamily. It encodes a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. This protein is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea. The protein is additionally expressed in keratinocytes, the pituitary gland, B-cells and T-cells.
Specifications
CHRNA9 | |
Polyclonal | |
Unconjugated | |
CHRNA9 | |
2410015I05Rik; acetylcholine receptor; acetylcholine receptor alpha 9 subunit (nAChR); acetylcholine receptor, neuronal nicotinic, alpha-9 subunit; Acra9; cholinergic receptor; cholinergic receptor nicotinic alpha 9 subunit; cholinergic receptor, nicotinic alpha 9; cholinergic receptor, nicotinic, alpha 9; cholinergic receptor, nicotinic, alpha 9 (neuronal); cholinergic receptor, nicotinic, alpha 9a; cholinergic receptor, nicotinic, alpha polypeptide 9; chrna9; chrna9a; EG666827; Gm8311; HSA243342; LOW QUALITY PROTEIN: neuronal acetylcholine receptor subunit alpha-9; NACHR alpha 9; NACHR alpha-9; NACHRA9; neuronal acetylcholine receptor protein, alpha-9 subunit; neuronal acetylcholine receptor subunit alpha-9; nicotinic acetylcholine receptor subunit alpha 9; nicotinic acetylcholine receptor subunit alpha-9; si:dkey-274k22.9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55584 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UGM1 | |
CHRNA9 | |
Synthetic peptide directed towards the N-terminal of human CHRNA9 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction