Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549341
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549341 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549341 Supplier Thermo Scientific Supplier No. PA549341
Add to Cart
Add to Cart

CHRNA9 Polyclonal Antibody

CHRNA9 Polyclonal Antibody for Western Blot, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA9
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CHRNA9
Gene Accession No. Q9UGM1
Gene Alias 2410015I05Rik; acetylcholine receptor; acetylcholine receptor alpha 9 subunit (nAChR); acetylcholine receptor, neuronal nicotinic, alpha-9 subunit; Acra9; cholinergic receptor; cholinergic receptor nicotinic alpha 9 subunit; cholinergic receptor, nicotinic alpha 9; cholinergic receptor, nicotinic, alpha 9; cholinergic receptor, nicotinic, alpha 9 (neuronal); cholinergic receptor, nicotinic, alpha 9a; cholinergic receptor, nicotinic, alpha polypeptide 9; chrna9; chrna9a; EG666827; Gm8311; HSA243342; LOW QUALITY PROTEIN: neuronal acetylcholine receptor subunit alpha-9; NACHR alpha 9; NACHR alpha-9; NACHRA9; neuronal acetylcholine receptor protein, alpha-9 subunit; neuronal acetylcholine receptor subunit alpha-9; nicotinic acetylcholine receptor subunit alpha 9; nicotinic acetylcholine receptor subunit alpha-9; si:dkey-274k22.9
Gene Symbols CHRNA9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 8-42 amino acids from the N-terminal region of human CHRNA9.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55584
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.