Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518924
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518924 Supplier Invitrogen™ Supplier No. PA518924
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNB1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Chrnb1
Gene Accession No. P11230
Gene Alias acetylcholine receptor; acetylcholine receptor beta; acetylcholine receptor subunit beta; acetylcholine receptor, nicotinic, beta 1 (muscle); AChR beta; Achr-2; ACHRB; Acrb; cholinergic receptor; cholinergic receptor nicotinic beta 1 subunit; cholinergic receptor nicotinic beta polypeptide 1; cholinergic receptor, nicotinic beta 1; cholinergic receptor, nicotinic, beta 1 (muscle); cholinergic receptor, nicotinic, beta polypeptide 1 (muscle); CHRNB; Chrnb 1; CHRNB1; CMS1D; CMS2A; CMS2C; nAChRB1; RNACRB1; SCCMS
Gene Symbols Chrnb1
Host Species Goat
Immunogen Synthetic peptide sequence (QEQEDHDALKED) corresponding to the internal amino acids of CHRNB1 (aa 448-460).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1140
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.