Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529991 Supplier Invitrogen™ Supplier No. PA529991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (88%), Rat (87%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Chrnb1
Gene Accession No. P11230
Gene Alias acetylcholine receptor; acetylcholine receptor beta; acetylcholine receptor subunit beta; acetylcholine receptor, nicotinic, beta 1 (muscle); AChR beta; Achr-2; ACHRB; Acrb; cholinergic receptor; cholinergic receptor nicotinic beta 1 subunit; cholinergic receptor nicotinic beta polypeptide 1; cholinergic receptor, nicotinic beta 1; cholinergic receptor, nicotinic, beta 1 (muscle); cholinergic receptor, nicotinic, beta polypeptide 1 (muscle); CHRNB; Chrnb 1; CHRNB1; CMS1D; CMS2A; CMS2C; nAChRB1; RNACRB1; SCCMS
Gene Symbols Chrnb1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AChR beta 1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.