Learn More
CHRNB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA552213
Description
Immunogen sequence: LSVVVLNLHH RSPHTHQMPL WVRQIFIHKL PLYLRLKRPK PERDLMPEPP HCSSPGSGWG RGTDEYFIRK PPSDFLFPKP NRFQPELSAP DLRRFIDGPN RAVALLPELR EVVSSISYIA RQLQEQEDHD ALKEDWQF Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.
Specifications
CHRNB1 | |
Polyclonal | |
Unconjugated | |
CHRNB1 | |
CHRNB1; ACHRB; cholinergic receptor, nicotinic, beta 1 (muscle) ; CHRNB; CMS1D; CMS2A; SCCMS; cholinergic receptor, nicotinic, beta polypeptide 1 (muscle); nicotinic acetylcholine receptor beta 1 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P11230 | |
CHRNB1 | |
Recombit protein corresponding to Human CHRNB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only