Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNB2 Monoclonal Antibody (2D12A2)
GREENER_CHOICE

Catalog No. PIMA531692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531692 Supplier Invitrogen™ Supplier No. MA531692
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31692 has been tested in indirect ELISA.

Neuronal acetylcholine receptors are homo- or heteropentameric complexes composed of homologous alpha and beta subunits. They belong to a superfamily of ligand-gated ion channels which allow the flow of sodium and potassium across the plasma membrane in response to ligands such as acetylcholine and nicotine. This gene encodes one of several beta subunits. Mutations in this gene are associated with autosomal dominant nocturnal frontal lobe epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNB2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2D12A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CHRNB2
Gene Accession No. P17787
Gene Alias [b]2-nAchR; acetylcholine receptor; acetylcholine receptor beta 2 neural; acetylcholine receptor, nicotinic, beta 2 (neuronal); Acrb2; Acrb-2; beta2 human neuronal nicotinic acetylcholine receptor; C030030P04Rik; cholinergic receptor; cholinergic receptor nicotinic beta 2 subunit; cholinergic receptor nicotinic beta polypeptide 2; cholinergic receptor, nicotinic beta 2; cholinergic receptor, nicotinic, beta 2 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal); cholinergic receptor, nicotinic, beta subunit 2; CHRNB2; EFNL3; nAChRB2; N-alpha 1; Neuronal acetylcholine receptor non-alpha-1 chain; neuronal acetylcholine receptor subunit beta-2; neuronal acetylcholine receptor, beta 2; neuronal nicotinic acetylcholine receptor; neuronal nicotinic acetylcholine receptor beta 2; nicotinic acetylcholine receptor beta 2 subunit
Gene Symbols CHRNB2
Host Species Mouse
Immunogen Purified recombinant fragment of human CHRNB2 (AA: extra 26-233) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1141
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.