Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNB2 Mouse anti-Human, Clone:2D12A2, Abnova™

Catalog No. 89334384 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-384 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-384 Supplier Abnova Corporation Supplier No. MAB17936
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human CHRNB2.

Neuronal acetylcholine receptors are homo- or heteropentameric complexes composed of homologous alpha and beta subunits. They belong to a superfamily of ligand-gated ion channels which allow the flow of sodium and potassium across the plasma membrane in response to ligands such as acetylcholine and nicotine. This gene encodes one of several beta subunits. Mutations in this gene are associated with autosomal dominant nocturnal frontal lobe epilepsy. [provided by RefSeq]

Specifications

Antigen CHRNB2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2D12A2
Conjugate Unconjugated
Description cholinergic receptor, nicotinic, beta 2 (neuronal)
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CHRNB2
Gene Alias EFNL3, nAChRB2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 26-233 of human CHRNB2.
Quantity 100 μg
Gene ID (Entrez) 1141
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.