Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHRNB2 Mouse anti-Human, Clone:2D12A2, Abnova™
Mouse monoclonal antibody raised against recombinant human CHRNB2.
Supplier: Abnova Corporation MAB17554
Description
Neuronal acetylcholine receptors are homo- or heteropentameric complexes composed of homologous alpha and beta subunits. They belong to a superfamily of ligand-gated ion channels which allow the flow of sodium and potassium across the plasma membrane in response to ligands such as acetylcholine and nicotine. This gene encodes one of several beta subunits. Mutations in this gene are associated with autosomal dominant nocturnal frontal lobe epilepsy. [provided by RefSeq]Specifications
CHRNB2 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry Flow Cytometry (1:200-1:400) Immunohistochemistry The optimal working dilution should be determined by the end user. |
|
CHRNB2 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Western Blot | |
2D12A2 | |
cholinergic receptor, nicotinic, beta 2 (neuronal) | |
In PBS (0.05% sodium azide) | |
EFNL3, nAChRB2 | |
Recombinant protein corresponding to amino acid 26-233 of human CHRNB2 from E. coli. | |
1141 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction