Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHRNB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529792 Supplier Invitrogen™ Supplier No. PA529792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (95%), Chicken (98%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Neuronal acetylcholine receptors are homo- or heteropentameric complexes composed of homologous alpha and beta subunits. They belong to a superfamily of ligand-gated ion channels which allow the flow of sodium and potassium across the plasma membrane in response to ligands such as acetylcholine and nicotine. This gene encodes one of several beta subunits. Mutations in this gene are associated with autosomal dominant nocturnal frontal lobe epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CHRNB2
Gene Accession No. P12390, P17787, Q9ERK7
Gene Alias [b]2-nAchR; acetylcholine receptor; acetylcholine receptor beta 2 neural; acetylcholine receptor, nicotinic, beta 2 (neuronal); Acrb2; Acrb-2; beta2 human neuronal nicotinic acetylcholine receptor; C030030P04Rik; cholinergic receptor; cholinergic receptor nicotinic beta 2 subunit; cholinergic receptor nicotinic beta polypeptide 2; cholinergic receptor, nicotinic beta 2; cholinergic receptor, nicotinic, beta 2 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal); cholinergic receptor, nicotinic, beta subunit 2; CHRNB2; EFNL3; nAChRB2; N-alpha 1; Neuronal acetylcholine receptor non-alpha-1 chain; neuronal acetylcholine receptor subunit beta-2; neuronal acetylcholine receptor, beta 2; neuronal nicotinic acetylcholine receptor; neuronal nicotinic acetylcholine receptor beta 2; nicotinic acetylcholine receptor beta 2 subunit
Gene Symbols CHRNB2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AChR beta 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1141, 11444, 54239
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.