Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHRNB2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518920
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Neuronal acetylcholine receptors are homo- or heteropentameric complexes composed of homologous alpha and beta subunits. They belong to a superfamily of ligand-gated ion channels which allow the flow of sodium and potassium across the plasma membrane in response to ligands such as acetylcholine and nicotine. This gene encodes one of several beta subunits. Mutations in this gene are associated with autosomal dominant nocturnal frontal lobe epilepsy.
Specifications
CHRNB2 | |
Polyclonal | |
Unconjugated | |
CHRNB2 | |
[b]2-nAchR; acetylcholine receptor; acetylcholine receptor beta 2 neural; acetylcholine receptor, nicotinic, beta 2 (neuronal); Acrb2; Acrb-2; beta2 human neuronal nicotinic acetylcholine receptor; C030030P04Rik; cholinergic receptor; cholinergic receptor nicotinic beta 2 subunit; cholinergic receptor nicotinic beta polypeptide 2; cholinergic receptor, nicotinic beta 2; cholinergic receptor, nicotinic, beta 2 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal); cholinergic receptor, nicotinic, beta subunit 2; CHRNB2; EFNL3; nAChRB2; N-alpha 1; Neuronal acetylcholine receptor non-alpha-1 chain; neuronal acetylcholine receptor subunit beta-2; neuronal acetylcholine receptor, beta 2; neuronal nicotinic acetylcholine receptor; neuronal nicotinic acetylcholine receptor beta 2; nicotinic acetylcholine receptor beta 2 subunit | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
1141 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P17787 | |
CHRNB2 | |
Synthetic peptide sequence (QPRHHCARQRLR) corresponding to the internal amino acids of CHRNB2 (aa 351-363). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction